missing translation for 'onlineSavingsMsg'
Learn More

Recoverin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16856223 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16856223 20 μL 20µL
16846223 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16856223

Brand: Proteintech 100731AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Calcium binding proteins are implicated as being a helper molecule in aberrant tissue growth of tumors. Recoverin is a neurologically important calcium binding protein, involved in tumor pathology. It is conserved across species and there are indications of several isoforms of this molecule. Recoverin antibodies are a useful tool for distinguishing between pineal gland-derived tissue and the surrounding tissue of the brain. They can also be used to accurately determine maligt tissue in this location of the brain. In pineal tumor tissue, the normal tissue staining pattern is lost, indicating the tissue′s loss of this protein.

Specifications

Antigen Recoverin
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RCVRN
Gene Accession No. P34057, P35243
Gene Alias Protein CAR, RCV1, RCVRN, recoverin
Gene Symbols RCVRN
Host Species Rabbit
Immunogen Recoverin Fusion Protein Ag0119
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140936, 19674, 5957
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.