missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Xenopus TGF-beta 1 / TGFB1 Protein

A DNA sequence encoding the Xenopus laevis (African clawed frog) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6781-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065800

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001081330.1
TGF-beta 1 / TGFB1 Protein
Research Use Only
TGFB1
Recombinant Protein
E. coli
GVGQEYCFGN NGPNCCVKPL YINFRKDLGW KWIHEPKGYE ANYCLGNCPY IWSMDTQYSK VLSLYNQNNP GASISPCCVP DVLEPLPIIY YVGRTAKVEQ LSNMVVRSCN CS
Greater than 90% as determined by SDS-PAGE.
397778
50 μg
Not determined.
Full Length
Xenopus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers