Learn More
enQuireBio™ Recombinant Xenopus Protein Wnt-8 Protein
A DNA sequence encoding the Xenopus laevis (African clawed frog) Protein Wnt-8, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP9601-ye-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16088501
  • 11060.00 NOK / 100µg
Estimated Shipment: 08-07-2024
Specifications
NP_001081637.1
Protein Wnt-8 Protein
Research Use Only
wnt8
Recombinant Protein
Yeast
AWSVNNFLMT GPKAYLTYSA SVAVGAQNGI EECKYQFAWE RWNCPESTLQ LATHNGLRSA TRETSFVHAI SSAGVMYTLT RNCSMGDFDN CGCDDSRNGR IGGRGWVWGG CSDNAEFGER ISKLFVDGLE TGQDARALMN LHNNEAGRLA VKETMKRTCK CHGISGSCSI QTCWLQLAEF RDIGNHLKIK HDQALKLEMD KRKMRSGNSA DNRGAIADAF SSVAGSELIF LEDSPDYCLK NISLGLQGTE GRECLQSGKN LSQWERRSCK RLCTDCGLRV EEKKTEIISS CNCKFHWCCT VKCEQCKQVV IKHFCARRER DSNMLNTKRK NRGHRR
Greater than 90% as determined by SDS-PAGE.
397970
100 μg
Not determined.
Full Length
Xenopus
His
Tris-based buffer, 50% glycerol