missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Foldase protein PrsA Protein
A DNA sequence encoding the Staphylococcus aureus (strain COL) Foldase protein PrsA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7670-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000782121.1 | |
10 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Foldase protein PrsA Protein | |
Research Use Only | |
prsA | |
Recombinant Protein | |
E. coli | |
CGASATDSKE NTLISSKAGD VTVADTMKKI GKDQIANASF TEMLNKILAD KYKNKVNDKK IDEQIEKMQK QYGGKDKFEK ALQQQGLTAD KYKENLRTAA YHKELLSDKI KISDSEIKED SKKASHILIK VKSKKSDKEG LDDKEAKQKA EEIQKEVSKD PSKFGEIAKK ESMDTGSAKK DGELGYVLKG QTDKDFEKAL FKLKDGEVSE VVKSSFGYHI IKADKPTDFN SEKQSLKEKL VDQKVQKNPK LLTDAYKDLL KEYDVDFKDR DIKSVVEDKI LNPEKLKQGG AQGGQSGMSQ | |
Greater than 90% as determined by SDS-PAGE. |