missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus fabD Protein

A DNA sequence encoding the Staphylococcus aureus (strain COL) fabD, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7663-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16080061

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000047348.1
50 μg
Not determined.
Full Length
S. aureus
His
Tris-based buffer, 50% glycerol
S. aureus fabD Protein
Research Use Only
fabD
Recombinant Protein
Yeast
MSKTAIIFPG QGAQKVGMAQ DLFNNNDQAT EILTSAANTL DFDILETMFT DEEGKLGETE NTQPALLTHS SALLAALKNL NPDFTMGHSL GEYSSLVAAD VLSFEDAVKI VRKRGQLMAQ AFPTGVGSMA AVLGLDFDKV DEICKSLSSD DKIIEPANIN CPGQIVVSGH KALIDELVEK GKSLGAKRVM PLAVSGPFHS SLMKVIEEDF SSYINQFEWR DAKFPVVQNV NAQGETDKEV IKSNMVKQLY SPVQFINSTE WLIDQGVDHF IEIGPGKVLS GLIKKINRDV KLTSIQTLED VKGWNEND
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers