missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat TNF-alpha Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
TNF alpha Protein is fully biologically active when compared to standard. The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 0.05 ng/mL, corresponding to a specific activity of > 2.0 x 10^7 IU/mg in the presence of actinomycin D.
Specifications
Specifications
| Accession Number | P16599, P16599, P16599 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.2, 150 mM NaCl. |
| Gene ID (Entrez) | 7124 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 17.2 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of TNF-alpha as determined by LAL method. |
| Gene Alias | APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alpha, TNF-alpha/TNFSF2, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Gene Symbol | TNF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction