missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Thrombopoietin / THPO Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Thrombopoietin / THPO, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8583-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_112395.1 | |
Thrombopoietin / THPO Protein | |
Research Use Only | |
THPO | |
Recombinant Protein | |
E. coli | |
SPVPPACDPR LLNKLLRDSY LLHSRLSQCP DVNPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPPQGRTTA HKDPSALFLS LQQLLRGKVR FLLLVEGPAL CVRRTLPTTA VPSRTSQLLT LNK | |
Greater than 90% as determined by SDS-PAGE. |
108348162 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |