Learn More
enQuireBio™ Recombinant Rat Sortilin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Sortilin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6716-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_113955.1 | |
Sortilin Protein | |
Research Use Only | |
Sort1 | |
Recombinant Protein | |
E. coli | |
CEENDYTTWL AHSTDPGDYK DGCILGYKEQ FLRLRKSSVC QNGRDYVVAK QPSICPCSLE DFLCDFGYFR PENASECVEQ PELKGHELEF CLYGKEEHLT TNGYRKIPGD RCQGGMNPAR EVKDLKKKCT SNFLNPKKQN SKSSS | |
Greater than 90% as determined by SDS-PAGE. |
83576 | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |