missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinE1 / PAI-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinE1 / PAI-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6673-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036752.1 | |
SerpinE1 / PAI-1 Protein | |
Research Use Only | |
Serpine1 | |
Recombinant Protein | |
E. coli | |
SPLPESHTAQ QATNFGVKVF QHVVQASKDR NVVFSPYGVS SVLAMLQLTT AGKTRQQIQD AMGFNISERG TAPALRKLSK ELMGSWNKNE ISTADAIFVQ RDLELVQGFM PHFFKLFRTT VKQVDFSEVE RARFIINDWV ERHTKGMISD LLAKGAVNEL TRLVLVNALY FNGQWKTPFL EASTHQRLFH KSDGSTISVP MMAQNNKFNY TEFTTPDGHE YDILELPYHG ETLSMFIAAP FEKDVPLSAI TNILDAELIR QWKSNMTRLP RLLILPKFSL ETEVDLRGPL EKLGMTDIFS STQADFTSLS DQEQLSVAQA LQKVKIEVNE SGTVASSSTA ILVSARMAPT EMVLDRSFLF VVRHNPTETI LFMGQLMEP | |
Greater than 90% as determined by SDS-PAGE. |
24617 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |