missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinB2 / PAI-2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinB2 / PAI-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6669-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_067728.1 | |
SerpinB2 / PAI-2 Protein | |
Research Use Only | |
Serpinb2 | |
Recombinant Protein | |
Yeast | |
MEELSMANTM FALNLLKQIE QSNSTQNIFI SPWSISSTLA IVFLGAQANT EEQMAKVLNF DKIGSYDLTP GNPENFHGCD FAQHIQRDNY PVAILQAQAR DKIHSAFSSL SSTINTPRLG DYLLESANKL FGEKSARFKE EYIQRCKKYY STEPEAVDFL ECANEARKKI NSWVKTQTKG EIPNLLPEGS VDEDTKMVLV NTIYFKGRWK TPFQKRLNGL YPFRVNLNES KPVQMMYLRE KLNIGYIKDL KTQILELPYI GNISMFLLLP DEIEDSSTGL EMLEREINFD NFNKWISKET LDEDDVLVYI PKFKLAQNYE LKPILQRMGM EDAFNKGKAD FSGMSESNDL FLSEVFHQAT VDVNEEGTVA AGGTGAVMTG RTGHGGPQFV ADHPFLFFIM NNITRTILFV GRFSSP | |
Greater than 90% as determined by SDS-PAGE. |
60325 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |