missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PKLR / PKRL Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PKLR / PKRL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6498-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036756.3 | |
PKLR / PKRL Protein | |
Research Use Only | |
Pklr | |
Recombinant Protein | |
Yeast | |
MSVQENTLPQ QLWPWIFRSQ KDLAKSALSG APGGPAGYLR RASVAQLTQE LGTAFFQQQQ LPAAMADTFL EHLCLLDIDS QPVAARSTSI IATIGPASRS VDRLKEMIKA GMNIARLNFS HGSHEYHAES IANIREATES FATSPLSYRP VAIALDTKGP EIRTGVLQGG PESEVEIVKG SQVLVTVDPK FQTRGDAKTV WVDYHNITRV VAVGGRIYID DGLISLVVQK IGPEGLVTEV EHGGILGSRK GVNLPNTEVD LPGLSEQDLL DLRFGVQHNV DIIFASFVRK ASDVLAVRDA LGPEGQNIKI ISKIENHEGV KKFDEILEVS DGIMVARGDL GIEIPAEKVF LAQKMMIGRC NLAGKPVVCA TQMLESMITK ARPTRAETSD VANAVLDGAD CIMLSGETAK GSFPVEAVMM QHAIAREAEA AVYHRQLFEE LRRAAPLSRD PTEVTAIGAV EASFKCCAAA IIVLTKTGRS AQLLSQYRPR AAVIAVTRSA QAARQVHLSR GVFPLLYREP PEAIWADDVD RRVQFGIESG KLRGFLRVGD LVIVVTGWRP GSGYTNIMRV LSVS | |
Greater than 90% as determined by SDS-PAGE. |
24651 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |