Learn More
enQuireBio™ Recombinant Rat Gremlin 1 / GREM1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Gremlin 1 / GREM1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6118-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_062155.1 | |
Gremlin 1 / GREM1 Protein | |
Research Use Only | |
Grem1 | |
Recombinant Protein | |
E. coli | |
KKKGSQGAIP PPDKAQHNDS EQTQSPPQPG SRTRGRGQGR GTAMPGEEVL ESSQEALHVT ERKYLKRDWC KTQPLKQTIH EEGCNSRTII NRFCYGQCNS FYIPRHIRKE EGSFQSCSFC KPKKFTTMMV TLNCPELQPP TKKKRVTRVK QCRCISIDLD | |
Greater than 90% as determined by SDS-PAGE. |
50566 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |