missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Glutathione S-transferase P Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Glutathione S-transferase P, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6127-ec-200ug
Additional Details : Weight : 0.01000kg
Spezifikation
NP_036709.1 | |
Glutathione S-transferase P Protein | |
Research Use Only | |
Gstp1 | |
Recombinant Protein | |
E. coli | |
PPYTIVYFPV RGRCEATRML LADQGQSWKE EVVTIDVWLQ GSLKSTCLYG QLPKFEDGDL TLYQSNAILR HLGRSLGLYG KDQKEAALVD MVNDGVEDLR CKYGTLIYTN YENGKDDYVK ALPGHLKPFE TLLSQNQGGK AFIVGNQISF ADYNLLDLLL VHQVLAPGCL DNFPLLSAYV ARLSARPKIK AFLSSPDHLN RPINGNGKQ | |
Greater than 90% as determined by SDS-PAGE. |
24426 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |