missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Ephrin-A1 / EFNA1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Ephrin-A1 / EFNA1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5964-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_446051.2 | |
Ephrin-A1 / EFNA1 Protein | |
Research Use Only | |
EFNA1 | |
Recombinant Protein | |
Yeast | |
ADRHIVFWNS SNPKFREEDY TVHVQLNDYL DIICPHYEDD SVADAAMERY TLYMVEHQEY VTCEPQSKDQ VRWKCNQPSA KHGPEKLSEK FQRFTPFTLG KEFKEGHSYY YISKPIYHQE TQCLKLKVTV NGKITHSPHA HANPQEKRLQ ADDPEVQVLH SIGHS | |
Greater than 90% as determined by SDS-PAGE. |
94268 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |