missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat beta-NGF / Beta-NGF Protein
A DNA sequence encoding the Rattus norvegicus (Rat) beta-NGF / Beta-NGF Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9305-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001263984.1 | |
beta-NGF / Beta-NGF Protein | |
Research Use Only | |
Ngf | |
Recombinant Protein | |
Yeast | |
SSTHPVFHMG EFSVCDSVSV WVGDKTTATD IKGKEVTVLG EVNINNSVFK QYFFETKCRA PNPVESGCRG IDSKHWNSYC TTTHTFVKAL TTDDKQAAWR FIRIDTACVC VLSRKAARRG | |
Greater than 90% as determined by SDS-PAGE. |
310738 | |
10 μg | |
Not determined. | |
Full. Length. of. Mature. Protein. | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |