missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat beta-NGF / Beta-NGF Protein

A DNA sequence encoding the Rattus norvegicus (Rat) beta-NGF / Beta-NGF Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9305-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008071

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001263984.1
beta-NGF / Beta-NGF Protein
Research Use Only
Ngf
Recombinant Protein
Yeast
SSTHPVFHMG EFSVCDSVSV WVGDKTTATD IKGKEVTVLG EVNINNSVFK QYFFETKCRA PNPVESGCRG IDSKHWNSYC TTTHTFVKAL TTDDKQAAWR FIRIDTACVC VLSRKAARRG
Greater than 90% as determined by SDS-PAGE.
310738
200 μg
Not determined.
Full. Length. of. Mature. Protein.
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers