missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV GlycoProtein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7425-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
RABV GlycoProtein | |
Research Use Only | |
G | |
Recombinant Protein | |
E. coli | |
KFPIYTIPDK LGPWSPIDIH HLSCPNNLVV EDEGCTNLSG FSYMELKVGY ISAIKVNGFT CTGVVTEAET YTNFVGYVTT TFKRKHFRPT PDACRAAYNW KMAGDPRYEE SLHNPYPDYH WLRTVKTTKE SLVIISPSVA DLDPYDKSLH SRVFPSGKCS GITISSTYCS TNHDYTIWMP ENPRLGTSCD IFTNSRGKRA SKGGKTCGFV DERGLYKSLK GACKLKLCGV LGLRLMDGTW VAMQTSDETK WCPPDQLVNL HDFRSDEIEH LVVEELVKKR EECLDALESI MATKSVSFRR LSHLRKLVPG FGKAYTIFNK TLMEADAHYK SVRTWNEIIP SKGCLRVGGR CHPHVNGVFF NGIILGPDGH VLIPEMQSSL LQQHMELLES SVIPLMHPLA DPSTVFKDGD EAEDFVEVHL PDVHKQISGV DLGLPSWGKY | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
RABV | |
His-SUMO | |
Tris-based buffer, 50% glycerol |