missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit IL17 / IL17A Protein

A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) IL17 / IL17A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6212-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16021620

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

XP_002714544.1
IL17 / IL17A Protein
Research Use Only
IL17A
Recombinant Protein
Yeast
VKNGIAMPRN PGCPNAEDKN FPQNVKVSLN ILNKSVNSRR PSDYYNRSTS PWTLHRNEDR ERYPSVIWEA KCRHLGCVNA EGNEDHHMNS VPIQQEILVL RRESQHCPHS FRLEKMLVAV GCTCVTPIIH HMA
Greater than 90% as determined by SDS-PAGE.
100339322
50 μg
Not determined.
Full Length
Rabbit
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers