Learn More
Invitrogen™ Recombinant Protein G-HRP
Product Code. 10426873
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10426873 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10426873 Supplier Invitrogen™ Supplier No. 101223
 Restricted Item
Estimated Shipment: 14-10-2026
Recombinant Protein G-HRP
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This protein G is conjugated with highly purified horseradish peroxidase (RZ > 3.0) and is in enyme conjugate stabilizer containing 0.1% proclin.
Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG
    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Conjugate HRP
    Quantity 1 mL
    Form Liquid
    Protein Subtype Protein G
    Content And Storage 2-8°C
    Shipping Condition Wet Ice