Learn More
Invitrogen™ Recombinant Protein G
Product Code. 10726713
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10726713 1 mg 1mg
10100633 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10726713 Supplier Invitrogen™ Supplier No. 101200
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Recombinant Protein G
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. It is supplied as a salt-free lyophilized powder.
Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG
    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Conjugate Unconjugated
    Quantity 1 mg
    Form Powder
    Protein Subtype Protein G
    Content And Storage 2-8°C
    Shipping Condition Wet Ice