Learn More
Novex™ Recombinant Protein G
Product Code. 10100633
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. Quantity unitSize
10100633 5 mg 5mg
10726713 1 mg 1mg
2 options
Product Code. 10100633
Brand: Novex™ 101201
Recombinant Protein G
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. It is supplied as a salt-free lyophilized powder.
Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG
    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Conjugate Unconjugated
    Quantity 5 mg
    Form Powder
    Protein Subtype Protein G
    Content And Storage 2-8°C
    Shipping Condition Wet Ice