Learn More
enQuireBio™ Recombinant P. aeruginosa PA-I galactophilic lectin Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) PA-I galactophilic lectin, was expressed in the hosts and tags indicated. option, ab
Brand:  enQuireBio™ QP6995-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16066940
  • 10805.00 NOK / 100µg
Estimated Shipment: 08-07-2024
Specifications
NP_251260.1
P. aeruginosa PA-I galactophilic lectin Protein
Research Use Only
lecA
Recombinant Protein
E. coli
AWKGEVLANN EAGQVTSIIY NPGDVITIVA AGWASYGPTQ KWGPQGDREH PDQGLICHDA FCGALVMKIG NSGTIPVNTG LFRWVAPNNV QGAITLIYND VPGTYGNNSG SFSVNIGKDQ S
Greater than 90% as determined by SDS-PAGE.
882335
100 μg
Not determined.
Full Length
P. aeruginosa
His
Tris-based buffer, 50% glycerol