Learn More
enQuireBio™ Recombinant Mouse Toll-like receptor 7 Protein
A DNA sequence encoding the Mus musculus (Mouse) Toll-like receptor 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6801-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001277684.1 | |
Toll-like receptor 7 Protein | |
Research Use Only | |
Tlr7 | |
Recombinant Protein | |
E. coli | |
FRWFPKTLPC EVKVNIPEAH VIVDCTDKHL TEIPEGIPTN TTNLTLTINH IPSISPDSFR RLNHLEEIDL RCNCVPVLLG SKANVCTKRL QIRPGSFSGL SDLKALYLDG NQLLEIPQDL PSSLHLLSLE ANNIFSITKE NLTELVNIET LYLGQNCYYR NPCNVSYSIE KDAFLVMRNL KVLSLKDNNV TAVPTTLPPN LLELYLYNNI IKKIQENDFN NLNELQVLDL SGNCPRCYNV PYPCTPCENN SPLQIHDNAF NSLTELKVLR LHSNSLQHVP PTWFKNMRNL QELDLSQNYL AREIEEAKFL HFLPNLVELD FS | |
Greater than 90% as determined by SDS-PAGE. |
170743 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |