Learn More
enQuireBio™ Recombinant Mouse SerpinG1 / C1 inhibitor / C1IN Protein
A DNA sequence encoding the Mus musculus (Mouse) SerpinG1 / C1 inhibitor / C1IN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6676-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033906.2 | |
SerpinG1 / C1 inhibitor / C1IN Protein | |
Research Use Only | |
SERPING1 | |
Recombinant Protein | |
E. coli | |
TQDPLEAQAK SRESFPERDD SWSPPEPTVL PSTWPTTSVA ITITNDTMGK VANESFSQHS QPAAQLPTDS PGQPPLNSSS QPSTASDLPT QATTEPFCPE PLAQCSDSDR DSSEAKLSEA LTDFSVKLYH AFSATKMAKT NMAFSPFSIA SLLTQVLLGA GDSTKSNLES ILSYPKDFAC VHQALKGFSS KGVTSVSQIF HSPDLAIRDT YVNASQSLYG SSPRVLGPDS AANLELINTW VAENTNHKIR KLLDSLPSDT CLVLLNAVYL SAKWKITFEP KKMMAPFFYK NSMIKVPMMS SVKYPVAQFD DHTLKAKVGQ LQLSHNLSFV IVVPVFPKHQ LKDVEKALNP TVFKAIMKKL ELSKFLPTYL TMPHIKVKSS QDMLSVMEKL EFFDFTYDLN LCGLTEDPDL QVSAMKHETV LELTESGVEA AAASAISFGR SLPIFEVQRP FLFLLWDQQH RFPVFMGRVY DPR | |
Greater than 90% as determined by SDS-PAGE. |
12258 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |