missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Serpin H1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Serpin H1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6677-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001104513.1
Serpin H1 Protein
Research Use Only
Serpinh1
Recombinant Protein
Yeast
AEVKKPLEAA APGTAEKLSS KATTLAERST GLAFSLYQAM AKDQAVENIL LSPLVVASSL GLVSLGGKAT TASQAKAVLS AEKLRDEEVH TGLGELLRSL SNSTARNVTW KLGSRLYGPS SVSFADDFVR SSKQHYNCEH SKINFRDKRS ALQSINEWAS QTTDGKLPEV TKDVERTDGA LLVNAMFFKP HWDEKFHHKM VDNRGFMVTR SYTVGVTMMH RTGLYNYYDD EKEKLQMVEM PLAHKLSSLI ILMPHHVEPL ERLEKLLTKE QLKAWMGKMQ KKAVAISLPK GVVEVTHDLQ KHLAGLGLTE AIDKNKADLS RMSGKKDLYL ASVFHATAFE WDTEGNPFDQ DIYGREELRS PKLFYADHPF IFLVRDNQSG SLLFIGRLVR PKGDKMRDEL
Greater than 90% as determined by SDS-PAGE.
12406
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers