missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serpin H1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serpin H1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6677-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001104513.1 | |
Serpin H1 Protein | |
Research Use Only | |
Serpinh1 | |
Recombinant Protein | |
Yeast | |
AEVKKPLEAA APGTAEKLSS KATTLAERST GLAFSLYQAM AKDQAVENIL LSPLVVASSL GLVSLGGKAT TASQAKAVLS AEKLRDEEVH TGLGELLRSL SNSTARNVTW KLGSRLYGPS SVSFADDFVR SSKQHYNCEH SKINFRDKRS ALQSINEWAS QTTDGKLPEV TKDVERTDGA LLVNAMFFKP HWDEKFHHKM VDNRGFMVTR SYTVGVTMMH RTGLYNYYDD EKEKLQMVEM PLAHKLSSLI ILMPHHVEPL ERLEKLLTKE QLKAWMGKMQ KKAVAISLPK GVVEVTHDLQ KHLAGLGLTE AIDKNKADLS RMSGKKDLYL ASVFHATAFE WDTEGNPFDQ DIYGREELRS PKLFYADHPF IFLVRDNQSG SLLFIGRLVR PKGDKMRDEL | |
Greater than 90% as determined by SDS-PAGE. |
12406 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |