missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Phosphoserine aminotransferase Protein
A DNA sequence encoding the Mus musculus (Mouse) Phosphoserine aminotransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8039-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_803155.1 | |
Phosphoserine aminotransferase Protein | |
Research Use Only | |
Psat1 | |
Recombinant Protein | |
E. coli | |
MEATKQVVNF GPGPAKLPHS VLLEIQKQLL DYRGLGISVL EMSHRSSDFA KIIGNTENLV RELLAVPNNY KVIFVQGGGS GQFSAVPLNL IGLKAGRSAD YVVTGAWSAK AAEEAKKFGT VNIVHPKLGS YTKIPDPSTW NLNPDASYVY FCANETVHGV EFDFVPDVKG AVLVCDMSSN FLSRPVDVSK FGVIFAGAQK NVGSAGVTVV IVRDDLLGFS LRECPSVLDY KVQAGNNSLY NTPPCFSIYV MGMVLEWIKN NGGAAAMEKL SSIKSQMIYE IIDNSQGFYV CPVERQNRSR MNIPFRIGNA KGDEALEKRF LDKAVELNMI SLKGHRSVGG IRASLYNAVT TEDVEKLAAF MKNFLEMHQL | |
Greater than 90% as determined by SDS-PAGE. |
107272 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |