missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Phosphoserine aminotransferase Protein

A DNA sequence encoding the Mus musculus (Mouse) Phosphoserine aminotransferase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8039-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002321

  • 34365.00 NOK / 1mg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_803155.1
Phosphoserine aminotransferase Protein
Research Use Only
Psat1
Recombinant Protein
E. coli
MEATKQVVNF GPGPAKLPHS VLLEIQKQLL DYRGLGISVL EMSHRSSDFA KIIGNTENLV RELLAVPNNY KVIFVQGGGS GQFSAVPLNL IGLKAGRSAD YVVTGAWSAK AAEEAKKFGT VNIVHPKLGS YTKIPDPSTW NLNPDASYVY FCANETVHGV EFDFVPDVKG AVLVCDMSSN FLSRPVDVSK FGVIFAGAQK NVGSAGVTVV IVRDDLLGFS LRECPSVLDY KVQAGNNSLY NTPPCFSIYV MGMVLEWIKN NGGAAAMEKL SSIKSQMIYE IIDNSQGFYV CPVERQNRSR MNIPFRIGNA KGDEALEKRF LDKAVELNMI SLKGHRSVGG IRASLYNAVT TEDVEKLAAF MKNFLEMHQL
Greater than 90% as determined by SDS-PAGE.
107272
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers