missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Pcsk5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Pcsk5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6474-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001177412.1 | |
Pcsk5 Protein | |
Research Use Only | |
Pcsk5 | |
Recombinant Protein | |
E. coli | |
DYDLSHAQST YFNDPKWPSM WYMHCSDNTH PCQSDMNIEG AWKRGYTGKN IVVTILDDGI ERTHPDLMQN YDALASCDVN GNDLDPMPRY DASNENKHGT RCAGEVAATA NNSHCTVGIA FNAKIGGVRM LDGDVTDMVE AKSVSYNPQH VHIYSASWGP DDDGKTVDGP APLTRQAFEN GVRMGRRGLG SVFVWASGNG GRSKDHCSCD GYTNSIYTIS ISSTAESGKK PWYLEECSST LATTYSSGES YDKKIITTDL RQRCTDNHTG TSASAPMAAG IIALALEANP FLTWRDVQHV IVRTSRAGHL NANDWKTNAA GFKVSHLYGF GLMDAE | |
Greater than 90% as determined by SDS-PAGE. |
18552 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |