missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Oncomodulin Protein

A DNA sequence encoding the Mus musculus (Mouse) Oncomodulin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6444-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023360

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_149028.2
Oncomodulin Protein
Research Use Only
Ocm
Recombinant Protein
Yeast
SITDILSADD IAAALQECQD PDTFEPQKFF QTSGLSKMSA SQLKDIFQFI DNDQSGYLDE DELKYFLQRF QSDARELTES ETKSLMDAAD NDGDGKIGAD EFQEMVHS
Greater than 90% as determined by SDS-PAGE.
18261
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers