missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nidogen-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nidogen-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6420-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023190

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_035047.2
Nidogen-1 Protein
Research Use Only
Nid1
Recombinant Protein
E. coli
SPQRVNGKVK GRIFVGSSQV PVVFENTDLH SYVVMNHGRS YTAISTIPET VGYSLLPLAP IGGIIGWMFA VEQDGFKNGF SITGGEFTRQ AEVTFLGHPG KLVLKQQFSG IDEHGHLTIS TELEGRVPQI PYGASVHIEP YTELYHYSSS VITSSSTREY TVMEPDQDGA APSHTHIYQW RQTITFQECA HDDARPALPS TQQLSVDSVF VLYNKEERIL RYALSNSIGP VRDGSPDA
Greater than 90% as determined by SDS-PAGE.
18073
200 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers