missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Heme oxygenase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Heme oxygenase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9180-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034572.1 | |
Heme oxygenase 1 Protein | |
Research Use Only | |
Hmox1 | |
Recombinant Protein | |
Yeast | |
MERPQPDSMP QDLSEALKEA TKEVHIQAEN AEFMKNFQKG QVSREGFKLV MASLYHIYTA LEEEIERNKQ NPVYAPLYFP EELHRRAALE QDMAFWYGPH WQEIIPCTPA TQHYVKRLHE VGRTHPELLV AHAYTRYLGD LSGGQVLKKI AQKAMALPSS GEGLAFFTFP NIDSPTKFKQ LYRARMNTLE MTPEVKHRVT EEAKTAFLLN IELFEELQVM LTEEHKDQSP SQMASLRQRP ASLVQDTAPA ETPRGKPQIS TSSSQTPLLQ WVLTLSFLLA TVAVGIYAM | |
Greater than 90% as determined by SDS-PAGE. |
15368 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |