missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Galectin-7 Protein

A DNA sequence encoding the Mus musculus (Mouse) Galectin-7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6299-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072280

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Galectin-7 Protein
Research Use Only
LGALS7
Recombinant Protein
Yeast
SATHHKTSLP QGVRVGTVMR IRGLVPDQAG RFHVNLLCGE EQGADAALHF NPRLDTSEVV FNTKQQGKWG REERGTGIPF QRGQPFEVLL IATEEGFKAV VGDDEYLHFH HRLPPARVRL VEVGGDVQLH SLNIF
Greater than 90% as determined by SDS-PAGE.
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers