Learn More
enQuireBio™ Recombinant Mouse Fibroblast growth factor 15 Protein
A DNA sequence encoding the Mus musculus (Mouse) Fibroblast growth factor 15, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7474-ec-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16028940
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
NP_032029.1
Fibroblast growth factor 15 Protein
Research Use Only
Fgf15
Recombinant Protein
E. coli
RPLAQQSQSV SDEDPLFLYG WGKITRLQYL YSAGPYVSNC FLRIRSDGSV DCEEDQNERN LLEFRAVALK TIAIKDVSSV RYLCMSADGK IYGLIRYSEE DCTFREEMDC LGYNQYRSMK HHLHIIFIQA KPREQLQDQK PSNFIPVFHR SFFETGDQLR SKMFSLPLES DSMDPFRMVE DVDHLVKSPS FQK
Greater than 90% as determined by SDS-PAGE.
14170
500 μg
Not determined.
Full Length of Mature Protein
Mouse
His-SUMO
Tris-based buffer, 50% glycerol