missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse FGF-10 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
FGF-10 protein is fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 10^6 IU/mg.
Specifications
Specifications
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, 600 mM NaCl, pH 7.4, 1 mM mercaptoethanol. |
| Gene ID (Entrez) | 2255 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 19.5 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of FGF-10 as determined by LAL method. |
| Gene Alias | FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF-2/FGF-10, produced by fibroblasts of urinary bladder lamina propria |
| Gene Symbol | FGF10 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction