Learn More
enQuireBio™ Recombinant Mouse CXCL4 / PF4 Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL4 / PF4 Protein, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6484-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16083630
  • 7745.00 NOK / 100µg
Estimated Shipment: 03-07-2024
Specifications
NP_064316.1
CXCL4 / PF4 Protein
Research Use Only
Pf4
Recombinant Protein
E. coli
VTSAGPEESD GDLSCVCVKT ISSGIHLKHI TSLEVIKAGR HCAVPQLIAT LKNGRKICLD RQAPLYKKVI KKILES
Greater than 90% as determined by SDS-PAGE.
56744
100 μg
Not determined.
Full Length of Mature Protein
Mouse
His-SUMO
Tris-based buffer, 50% glycerol