Learn More
enQuireBio™ Recombinant Mouse CXCL4 / PF4 Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL4 / PF4 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6484-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_064316.1 | |
CXCL4 / PF4 Protein | |
Research Use Only | |
Pf4 | |
Recombinant Protein | |
E. coli | |
VTSAGPEESD GDLSCVCVKT ISSGIHLKHI TSLEVIKAGR HCAVPQLIAT LKNGRKICLD RQAPLYKKVI KKILES | |
Greater than 90% as determined by SDS-PAGE. |
56744 | |
500 μg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |