missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CHST4 Protein
Description
Measured by its ability to transfer sulfate from PAPS to N-acetyl-D-glucosamine. The specific activity is >35 pmol/min/μg, measured under the described conditions.
Specifications
Specifications
| Accession Number | Q9R1I1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
| Gene ID (Entrez) | 10164 |
| Molecular Weight (g/mol) | M.W. (Observed): 40-60 kDa, reducing conditions; M.W. (theoretical): 42 kDa |
| Name | Carbohydrate Sulfotransferase 4/CHST4 |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived mouse Carbohydrate Sulfotransferase 4/CHST4 protein Arg30-Gly388 with an N-terminal 6-His tag Accession # Q9R1I1 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?