missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CCL3 / Mip1a Protein

A DNA sequence encoding the Mus musculus (Mouse) CCL3 / Mip1a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8644-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075731

  • 34365.00 NOK / 1mg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_035467.1
CCL3 / Mip1a Protein
Research Use Only
Mip1a
Recombinant Protein
E. coli
APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA
Greater than 90% as determined by SDS-PAGE.
20302
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers