missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ATP synthase subunit beta, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) ATP synthase subunit beta, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5694-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_058054.2 | |
ATP synthase subunit beta, mitochondrial Protein | |
Research Use Only | |
Atp5b | |
Recombinant Protein | |
E. coli | |
AAQASAAPKA GTATGRIVAV IGAVVDVQFD EGLPPILNAL EVQGRDSRLV LEVAQHLGES TVRTIAMDGT EGLVRGQKVL DSGAPIKIPV GPETLGRIMN VIGEPIDERG PIKTKQFAPI HAEAPEFIEM SVEQEILVTG IKVVDLLAPY AKGGKIGLFG GAGVGKTVLI MELINNVAKA HGGYSVFAGV GERTREGNDL YHEMIESGVI NLKDATSKVA LVYGQMNEPP GARARVALTG LTVAEYFRDQ EGQDVLLFID NIFRFTQAGS EVSALLGRIP SAVGYQPTLA TDMGTMQERI TTTKKGSITS VQAIYVPADD LTDPAPATTF AHLDATTVLS RAIAELGIYP AVDPLDSTSR IMDPNIVGNE HYDVARGVQK ILQDYKSLQD IIAILGMDEL SEEDKLTVSR ARKIQRFLSQ PFQVAEVFTG HMGKLVPLKE TIKGFQQILA GEYDHLPEQA FYMVGPIEEA VAKADKLAEE HGS | |
Greater than 90% as determined by SDS-PAGE. |
11947 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |