missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Alkaline Phosphatase / ALPL Protein
A DNA sequence encoding the Mus musculus (Mouse) Alkaline Phosphatase / ALPL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8885-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001274101.1 | |
Alkaline Phosphatase / ALPL Protein | |
Research Use Only | |
Alpl | |
Recombinant Protein | |
E. coli | |
PEKERDPSYW RQQAQETLKN ALKLQKLNTN VAKNVIMFLG DGMGVSTVTA ARILKGQLHH NTGEETRLEM DKFPFVALSK TYNTNAQVPD SAGTATAYLC GVKANEGTVG VSAATERTRC NTTQGNEVTS ILRWAKDAGK SVGIVTTTRV NHATPSAAYA HSADRDWYSD NEMPPEALSQ GCKDIAYQLM HNIKDIDVIM GGGRKYMYPK NRTDVEYELD EKARGTRLDG LDLISIWKSF KPRHKHSHYV WNRTELLALD PSRVDYLLGL FEPGDMQYEL NRNNLTDPSL SEMVEVALRI LTKNLKGFFL LVEGGRIDHG HHEGKAKQAL HEAVEMDQAI GKAGAMTSQK DTLTVVTADH SHVFTFGGYT PRGNSIFGLA PMVSDTDKKP FTAILYGNGP GYKVVDGERE NVSMVDYAHN NYQAQSAVPL RHETHGGEDV AVFAKGPMAH LLHGVHEQNY IPHVMAYASC IGANLDHCAW AGSG | |
Greater than 90% as determined by SDS-PAGE. |
11647 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |