Learn More
enQuireBio™ Recombinant Mouse Acid ceramidase Protein
A DNA sequence encoding the Mus musculus (Mouse) Acid ceramidase, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8262-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16003621
  • 7745.00 NOK / 100µg
Estimated Shipment: 02-07-2024
Specifications
NP_062708.1
Acid ceramidase Protein
Research Use Only
ASAH1
Recombinant Protein
E. coli
QAQDVPPWTE DCRKSTYPPS GPTYRGPVPW HTINLDLPPY KRWHELLAQK APALRILVNS ITSLVNTFVP SGKLMKMVDQ KLPGMIGSLP DPFGEEMRGI ADVTGIPLGE IISFNIFYEL FTM
Greater than 80% as determined by SDS-PAGE.
11886
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His-GST
Tris-based buffer, 50% glycerol