missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 592 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 592, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8453-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055445.2 | |
Zinc finger protein 592 Protein | |
Research Use Only | |
ZNF592 | |
Recombinant Protein | |
E. coli | |
MGDMKTPDFD DLLAAFDIPD PTSLDAKEAI QTPSEENESP LKPPGICMDE SVSLSHSGSA PDVPAVSVIV KNTSRQESFE AEKDHITPSL LHNGFRGSDL PPDPHNCGKF DSTFMNGDSA RSFPGKLEPP KSEPLPTFNQ FSPISSPEPE DPIKDNGFGI KPKHSDSYFP PPLGCGAVGG PVLEALAKFP VPELHMFDHF CKKEPKPEPL PLGSQQEHEQ SGQNTVEPHK DPDATRFFGE AL | |
Greater than 90% as determined by SDS-PAGE. |
9640 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |