missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Very long-chain acyl-CoA synthetase Protein
A DNA sequence encoding the Homo sapiens (Human) Very long-chain acyl-CoA synthetase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6694-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001153101.1 | |
Very long-chain acyl-CoA synthetase Protein | |
Research Use Only | |
SLC27A2 | |
Recombinant Protein | |
Yeast | |
GATLALRTKF SASQFWDDCR KYNVTVIQYI GELLRYLCNS PQKPNDRDHK VRLALGNGLR GDVWRQFVKR FGDICIYEFY AATEGNIGFM NYARKVGAVG RVNYLQKKII TYDLIKYDVE KDEPVRDENG YCVRVPKGEV GLLVCKITQL TPFNGYAGAK AQTEKKKLRD VFKKGDLYFN SGDLLMVDHE NFIYFHDRVG DTFRWKGENV ATTEVADTVG LVDFVQEVNV YGVHVPDHEG RIGMASIKMK ENHEFDGKKL FQHIADYLPS YARPRFLRIQ DTIEITGTFK HRKMTLVEEG FNPAVIKDAL YFLDDTAKMY VPMTEDIYNA ISAKTLKL | |
Greater than 90% as determined by SDS-PAGE. |
11001 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |