missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGFR2 / Flk-1 / CD309 / KDR Protein
A DNA sequence encoding the Homo sapiens (Human) VEGFR2 / Flk-1 / CD309 / KDR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6258-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002244.1 | |
VEGFR2 / Flk-1 / CD309 / KDR Protein | |
Research Use Only | |
KDR | |
Recombinant Protein | |
E. coli | |
ASVGLPSVSL DLPRLSIQKD ILTIKANTTL QITCRGQRDL DWLWPNNQSG SEQRVEVTEC SDGLFCKTLT IPKVIGNDTG AYKCFYRETD LASVIYVYVQ DYRSPFIASV SDQHGVVYIT ENKNKTVVIP CLGSISNLNV SLCARYPEKR FVPDGNRISW DSKKGFTIPS YMISYAGMVF CEAKINDESY QSIMYIVVVV GYRIYDVVLS PSHGIELSVG EKLVLNCTAR TELNVGIDFN WEYPSSKHQH KKLVNRDLKT QSGSEMKKFL STLTIDGVTR SDQGLYTCAA SSGLMTKKNS TFVRVHEKPF VAFGSGMESL VEATVGERVR IPAKYLGYPP PEIKWYKNGI PLESNHTIKA GHVLTIMEVS ERDTGNYTVI LTNPISKEKQ SHVVSLVVYV PPQIGEKSLI SPVDSYQYGT TQTLTCTVYA IPPPHHIHWY WQLEEECANE PSQAVSVTNP YPCEEWRSVE DFQGGNKIEV NKNQFALIEG KNKTVSTLVI QAANVSALYK CEAVNKVGRG ERVISFHVTR GPEITLQPDM QPTEQESVSL WCTADRSTFE NLTWYKLGPQ PLPIHVGELP TPVCKNLDTL WKLNATMFSN STNDILIMEL KNASLQDQGD YVCLAQDRKT KKRHCVVRQL TVLERVAPTI TGNLENQTTS IGESIEVSCT ASGNPPPQIM WFKDNETLVE DSGIVLKDGN RNLTIRRVRK EDEGLYTCQA CSVLGCAKVE AFFIIEGAQE KTNLE | |
Greater than 90% as determined by SDS-PAGE. |
3791 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |