Learn More
R&D Systems™ Recombinant Human VEGF 111 Protein
Description
The Recombinant Human VEGF 111 Protein is derived from E. coli. The Recombinant Human VEGF 111 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 7422 |
| Molecular Weight (g/mol) | 13 kDa |
| Name | VEGF 111 |
| Quantity | 10 μg |
| Source | E. coli-derived human VEGF protein Ala27-Arg137,with an N-terminal Met & Pro28-Arg137 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |