missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C20orf197 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C20orf197, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7850-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001289742.1 | |
Uncharacterized protein C20orf197 Protein | |
Research Use Only | |
C20orf197 | |
Recombinant Protein | |
E. coli | |
MVALFQHSPY WQADGYGHSH RLKCQHFKQH RQYNDKLEIS SNLGPQFNAL LNILLNIVHP TLSHDTRRSK GLKIEGLLQS RELGNSWTVT MCIWVLKALQ SSAPNKPLDW LDPMPCFQNL LARGTP | |
Greater than 90% as determined by SDS-PAGE. |
284756 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |