missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Ubiquitin Protein
Click to view available options
:
10mg; Carrier Free
Description
The Recombinant Human Ubiquitin Protein is derived from E. coli. The Recombinant Human Ubiquitin Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Accession Number | P0CG47.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in deionized water. |
| Gene ID (Entrez) | 7314 |
| Molecular Weight (g/mol) | 8.6 kDa |
| Name | Ubiquitin |
| Quantity | 10mg |
| Source | E. coli-derived human Ubiquitin protein Met1 - Gly76 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction