missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-like-conjugating enzyme ATG3 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like-conjugating enzyme ATG3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5690-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265641.1 | |
Ubiquitin-like-conjugating enzyme ATG3 Protein | |
Research Use Only | |
ATG3 | |
Recombinant Protein | |
E. coli | |
MQNVINTVKG KALEVAEYLT PVLKESKFKE TGVITPEEFV AAGDHLVHHC PTWQWATGEE LKVKAYLPTG KQFLVTKNVP CYKRCKQMEY SDELEAIIEE DDGDGGWVDT YHNTGITGIT EAVKEITLEN KDNIRLQDCS ALCEEEEDED EGEAADMEEY EESGLLETDE ATLDTRKIVE ACKAKTDAGG EDAILQTRTY DLYITYDKYY QTPRLWLFGY DEQRQPLTVE HMYEDISQDH VKKTVTIENH PHLPPPPMCS VHPCRHAEVM KKIIETVAEG GGELGVHMYL LIFLKFVQAV IPTIEYDYTR HFTM | |
Greater than 90% as determined by SDS-PAGE. |
64422 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |