missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tyrosine-protein kinase JAK2 Protein
A DNA sequence encoding the Homo sapiens (Human) Tyrosine-protein kinase JAK2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6246-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001309123.1 | |
Tyrosine-protein kinase JAK2 Protein | |
Research Use Only | |
JAK2 | |
Recombinant Protein | |
E. coli | |
KPLSALDSQR KLQFYEDRHQ LPAPKWAELA NLINNCMDYE PDFRPSFRAI IRDLNSLFTP DYELLTENDM LPNMRIGALG FSGAFEDRDP TQFEERHLKF LQQLGKGNFG SVEMCRYDPL QDNTGEVVAV KKLQHSTEEH LRDFEREIEI LKSLQHDNIV KYKGVCYSAG RRNLKLIMEY LPYGSLRDYL QKHKERIDHI KLLQYTSQIC KGMEYLGTKR YIHRDLATRN ILVENENRVK IGDFGLTKVL PQDKEYYKVK EPGESPIFWY APESLTESKF SVASDVWSFG VVLYELFTYI EKSKSPPAEF MRMIGNDKQG QMIVFHLIEL LKNNGRLPRP DGCPDEIYMI MTECWNNNVN QRPSFRDLAL RVDQIRDNMA G | |
Greater than 90% as determined by SDS-PAGE. |
3717 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |